missing translation for 'onlineSavingsMsg'
Learn More

CD13 Mouse anti-Human, Clone: WM-15 (WM15), eBioscience™

Mouse Monoclonal Antibody

442.00 SEK - 892.00 SEK

Specifications

Antigen CD13
Clone WM-15 (WM15)
Target Species Human
Conjugate Unconjugated
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15236817
missing translation for 'viewDocuments'
Affymetrix eBioscience
14-0138-80
25 μg
442.00 SEK
25µg
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment
07-01-2021
Add to basket
 
15246817
missing translation for 'viewDocuments'
Affymetrix eBioscience
14-0138-82
100 μg
892.00 SEK
100µg
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment
07-01-2021
Add to basket
 
Description

Description

CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.
Specifications

Specifications

CD13
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C
WM-15 (WM15)
Unconjugated
Flow Cytometry, Immunohistochemistry
Primary
Monoclonal
PBS with 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.