missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
CD133 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44249V
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
CD133 Polyclonal specifically detects CD133 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifica
| CD133 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PROM1 | |
| A synthetic peptide made to an internal portion of the mouse CD133 protein (between residues 300-400) [Uniprot: O54990] | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| Mouse | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto