missing translation for 'onlineSavingsMsg'
Learn More

CD138 (Syndecan-1) Mouse anti-Human, PE, Clone: DL-101, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1389-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15576886

  • 900.00 SEK / 25 tests


Estimated Shipment
04-01-2021
Add to basket

Description

Description

CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.
Specifications

Specifications

CD138 (Syndecan-1)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P18827
CD antigen 138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; SDC; SDC1; syndecan proteoglycan 1; syndecan-1; synstatin
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
DL-101
PE
SDC1
Liquid
SDC1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
6382
Videos
SDS
Documents

For Research Use Only.