missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Monoclonal Antibody (TuK4), Pacific Orange™, Invitrogen™
Mouse Monoclonal Antibody
2540.00 SEK - 4985.00 SEK
Specifications
Antigen | CD14 |
---|---|
Clone | TuK4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Pacific Orange |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10335633
|
Invitrogen
MHCD1430 |
500 μL |
4985.00 SEK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11340782
|
Invitrogen
MHCD1430TR |
25 Tests |
2540.00 SEK
Each |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Our Pacific Orange™ dye is optimally excited by the violet laser, and it is recommended that a 575, 585 or 600 nm band pass filter be used for optimal detection. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Flow Cytometry | |
Pacific Orange | |
Mouse | |
PBS with BSA and 0.1% sodium azide | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Cd14 | |
IgG2a | |
purified | |
Antibody |
TuK4 | |
Monoclonal | |
Liquid | |
Human | |
P08571 | |
929 | |
Human CD14 | |
Primary | |
4° C, store in dark | |
Cd14 |