Learn More
CD14 Monoclonal Antibody (61D3), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
3110.00 SEK
Specifications
Antigen | CD14 |
---|---|
Clone | 61D3 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15510417
|
Affymetrix eBioscience
61-0149-42 |
100 Tests |
3110.00 SEK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 61D3 monoclonal antibody reacts with human CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14. Applications Reported: This 61D3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 61D3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P08571 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
61D3 | |
Flow Cytometry | |
PE-eFluor 610 | |
Mouse | |
Human | |
929 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.