Learn More
CD140a (PDGFRA) Monoclonal Antibody (APA5), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1378.00 SEK - 3450.00 SEK
Specifications
Antigen | CD140a (PDGFRA) |
---|---|
Clone | APA5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933754
|
Affymetrix eBioscience
67-1401-82 |
100 μg |
3450.00 SEK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923754
|
Affymetrix eBioscience
67-1401-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This FLEG monoclonal antibody recognizes human CD162, which is also known as P-selectin glyocoprotein ligand-1 (PSGL-1). This 120-kDa protein exists as a homodimer on the surface of monocytes, neutrophils, granulocytes, peripheral T cells, some B cells, and a subset of CD34+ hematopoietic progenitor cells (HPCs) in the bone marrow. CD162 binds CD62P (P-selectin), CD62E (E-selectin), and CD62L (L-selectin) to mediate leukocyte interactions with each other as well as with activated platelets and endothelium. Thus, CD162 plays a critical role in leukocyte adhesion and rolling. Furthermore, the binding of CD162 to P-selectin on HPCs has been demonstrated to inhibit hematopoiesis. Studies have also demonstrated CD162 as the receptor for enterovirus 71. Applications Reported: This FLEG antibody has been reported for use in flow cytometric analysis. Applications Tested: This FLEG antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter.
CD140a (PDGFRA) Monoclonal antibody specifically detects CD140a (PDGFRA) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD140a (PDGFRA) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P26618 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
APA5 | |
Flow Cytometry | |
Super Bright 702 | |
Rat | |
Mouse | |
18595 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |