missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD140b (PDGFRB) Monoclonal Antibody (APB5), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1834.00 SEK - 4550.00 SEK
Specifications
Antigen | CD140b (PDGFRB) |
---|---|
Clone | APB5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16606324
|
Affymetrix eBioscience
25-1402-80 |
25 μg |
1834.00 SEK
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16616324
|
Affymetrix eBioscience
25-1402-82 |
100 μg |
4550.00 SEK
Each |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD140b (PDGFRB) Monoclonal antibody specifically detects CD140b (PDGFRB) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD140b (PDGFRB) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI528809; Beta platelet-derived growth factor receptor; beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140B; CD140b antigen; IBGC4; IMF1; JTK12; KOGS; PDGF beta chain; PDGF Receptor beta; Pdgfr; Pdgfr1; PDGFR-1; PDGFRB; PDGF-R-beta; PDGFR-beta; PENTT; platelet derived growth factor receptor beta; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide | |
Pdgfrb | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Pdgfrb |
APB5 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
P05622 | |
18596 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |