Läs mer
CD141, clone: LS17-9, eBioscience™
Beskrivning
Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
Specifikationer
Specifikationer
| Antigen | CD141 |
| Användningsområden | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Klassificering | Monoclonal |
| Klona | LS17-9 |
| Koncentration | 0.5mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 0.09% sodium azide; pH 7.2 |
| Gen | Thbd |
| Genaccessionsnr. | P15306 |
| Gene Alias | AHUS6, BDCA3, CD141, THPH12, THRM, TM, CD141 antigen, fetomodulin, DC141, THBD |
| Visa mer |
For Research Use Only.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.