missing translation for 'onlineSavingsMsg'
Learn More

CD144 (VE-cadherin) Mouse anti-Human, Alexa Fluor 700, Clone: 16B1, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-1449-42

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16606444

  • 3830.00 SEK / 100 tests
Estimated Shipment: 18-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling. Applications Reported: This 16B1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 16B1 antibody has been pre-diluted and tested by flow cytometric analysis of Human Umbilical Vein Endothelial Cells (HUVEC). This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor 700 emits at 723 nm and can be excited with the red laser (633-647 nm). Most instruments will require a 685 LP mirror and 710/20 band pass filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser

VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.
Specifications

Specifications

CD144 (VE-cadherin)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P33151
CDH5
Affinity chromatography
RUO
1003
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
16B1
Alexa Fluor 700
CDH5
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; BOS_17039; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); Cadherin-5; CD144; cd144 antigen; CDH5; endothelial-specific cadherin; Vascular endothelial cadherin; Vec; VEcad; VE-Cad; VE-cadherin; VECD
Mouse
100 Tests
Primary
Human
Antibody
IgG1
Videos
SDS
Documents
Certificates
Special Offers

Special Offers