missing translation for 'onlineSavingsMsg'
Learn More

CD144 (VE-cadherin) Mouse anti-Human, Biotin, Clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

900.00 SEK - 1968.00 SEK

Specifications

Antigen CD144 (VE-Cadherin)
Clone 16B1
Target Species Human
Conjugate Biotin
Isotype IgG1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15299729
View Documents
Affymetrix eBioscience
13-1449-80
25 μg
900.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment
07-01-2021
Add to basket
 
15209739
View Documents
Affymetrix eBioscience
13-1449-82
100 μg
1968.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment
04-12-2020
Add to basket
 
Description

Description

This gene is a classical cadherin from the cadherin superfamily and is located in a six-cadherin cluster in a region on the long arm of chromosome 16 that is involved in loss of heterozygosity events in breast and prostate cancer. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, the protein may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but its full length sequence has not been determined.
Specifications

Specifications

CD144 (VE-Cadherin)
Human
IgG1
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
16B1
Biotin
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Primary
Monoclonal
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.