Learn More
SSEA1 Monoclonal Antibody (VIMC6), Pacific Blue™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1528
Additional Details : Weight : 0.00050kg
Description
Our Pacific Blue™ dye is optimally excited by the violet laser and conjugates of this dye are strongly fluorescent even at neutral pH. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
SSEA1 Monoclonal antibody specifically detects SSEA1 in Human samples. It is validated for Flow Cytometry.Specifications
SSEA1, CD15 | |
Monoclonal | |
Pacific Blue | |
FUT4 | |
stage-specific embryonic antigen 1, CD15, SSEA-1, SSEA1, ELFT, FCT3A, FUC-TIV, FUTIV, LeX | |
Mouse | |
Affinity chromatography | |
ASR, ASR/IVD | |
2526 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
VIMC6 | |
PBS with BSA and 0.1% sodium azide | |
P22083 | |
FUT4 | |
Human CD15 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgM |