Learn More
SSEA1 Monoclonal Antibody (VIMC6), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1504
2160.10 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
SSEA1 Monoclonal antibody specifically detects SSEA1 in Human samples. It is validated for Flow Cytometry.Specifications
SSEA1, CD15 | |
Monoclonal | |
PE | |
FUT4 | |
stage-specific embryonic antigen 1, CD15, SSEA-1, SSEA1, ELFT, FCT3A, FUC-TIV, FUTIV, LeX | |
Mouse | |
purified | |
ASR, ASR/IVD | |
2526 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
VIMC6 | |
PBS with BSA and 0.1% sodium azide | |
P22083 | |
FUT4 | |
Human CD15 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgM |