missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ CD154 (CD40 Ligand) Monoclonal Antibody (MR1), PE-eFluor™ 610, eBioscience™, Invitrogen™

Produktkod. 16636464
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16636464 25 μg
16646464 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16636464 Supplier Invitrogen™ Supplier No. 61154180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154 (CD40 Ligand)
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona MR1
Koncentration 0.2 mg/mL
Konjugera PE-eFluor 610
Formulering PBS with 0.09% sodium azide; pH 7.2
Gen CD40LG
Genaccessionsnr. P27548
Gene Alias CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5
Gensymboler CD40LG
Värd art Armenian Hamster
Reningsmetod Affinity chromatography
Kvantitet 25 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 21947
Målarter Mouse
Innehåll och lagring 4°C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.