Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (24-31), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
954.00 SEK - 2750.00 SEK
Specifications
Antigen | CD154 (CD40 Ligand) |
---|---|
Clone | 24-31 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15506506
|
Affymetrix eBioscience
11-1548-42 |
100 Tests |
2750.00 SEK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15596496
|
Affymetrix eBioscience
11-1548-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 24-31 monoclonal antibody reacts with human CD154, a 39 kDa transmembrane glycoprotein also known as gp39 and CD40 ligand (CD40L). CD154 is a member of the TNF superfamily and is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells including B cells, monocytes/macrophages and dendritic cells, CD154 serves a crucial function in T-APC cognate interaction. CD154 interaction with CD40 transduces signals for T-dependent B-cell activation and induces B cell cycle entry. 24-31 cross-reacts with the cynomolgus monkey CD154. Applications Reported: 24-31 has been reported for use in flow cytometric analysis. Applications Tested: This 24-31 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.Specifications
CD154 (CD40 Ligand) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P29965 | |
CD40LG | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD40LG |
24-31 | |
Flow Cytometry | |
FITC | |
Mouse | |
Cynomolgus Monkey, Human | |
102138630, 959 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.