missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD157 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16337-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
BST1 (CD157, Bone marrow stromal cell antigen-1) is a stromal cell line-derived glycosylphosphatidylinositol-anchored molecule that facilitates pre-B-cell growth. The deduced amino acid sequence exhibits 33% similarity with CD38. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis. The polyclonal B-cell abnormalities in rheumatoid arthritis may be, at least in part, attributed to BST1 overexpression in the stromal cell population.Specifications
CD157 | |
Polyclonal | |
Unconjugated | |
BST1 | |
ADP ribosyl cyclase 2, Bone marrow stromal antigen 1, BST 1, BST1, cADPr hydrolase 2, CD157, Cyclic ADP ribose hydrolase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12182, 683, 81506 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q10588, Q63072, Q64277 | |
Bst1 | |
CD157 Fusion Protein Ag9491 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |