Learn More
CD158a/h/g Monoclonal Antibody (HP-MA4), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-1589-42
2043.43 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.23750kg
Description
Description: The monoclonal antibody HP-MA4 recognizes several CD158 proteins, also known as KIR2D, specifically KIR2DL1 (CD158a), KIR2DS1 (CD158h), KIR2DS3, and KIR2DS5 (CD158g). It does not recognize KIR2DL2 (CD158b1), KIR2DS2 (CD158j), KIR2DL4 (CD158d) or KIR2DS4 (CD158i) and crossreactivity to KIR2DL3 (CD158b2) and KIR2DL5 (CD158f) are unknown. CD158 proteins are members of the killer cell immunoglobulin-like receptors (KIR) family of transmembrane glycoproteins that are expressed on NK cells and a subset of T cells. Each KIR has an isoform with either a short (S) or long (L) cytoplasmic domain which transduces either an activating or inhibitory signal, respectively. The crossreactivity of the HP-MA4 antibody to KIR2DS3 and KIR2DS5 was published in 2013. Applications Reported: This HP-MA4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HP-MA4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD158a/h/g Monoclonal antibody specifically detects CD158a/h/g in Human samples. It is validated for Flow Cytometry.Specifications
CD158a/h/g | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P43626, P43631, P43632, Q14953, Q14954 | |
KIR2DL1, KIR2DS1, KIR2DS2, KIR2DS4, KIR2DS5 | |
Affinity chromatography | |
RUO | |
100132285, 3802, 3806, 3809, 3810 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HP-MA4 | |
PE-Cyanine7 | |
KIR2DL1 | |
183ActI; CD158; CD158 antigen-like family member A; CD158 antigen-like family member G; CD158 antigen-like family member H; CD158 antigen-like family member I; CD158 antigen-like family member J; CD158A; CD158b; CD158B1; CD158G; CD158H; CD158I; CD158j; cl-49; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5; killer cell immunoglobulin-like receptor 2DL1; Killer cell immunoglobulin-like receptor 2DS1; killer cell immunoglobulin-like receptor 2DS2; Killer cell immunoglobulin-like receptor 2DS4; killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor KIR2DS2; killer cell immunoglo | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
For Research Use Only.