missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD162 Monoclonal Antibody (TC2), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15720
Additional Details : Weight : 0.00280kg
Description
SELPLG is the high affinity counter-receptor for P-selectin on myeloid cells and stimulated T lymphocytes. As such, it plays a critical role in the tethering of these cells to activated platelets or endothelia expressing P-selectin. The organization of the SELPG gene closely resembles that of CD43 and the human platelet glycoprotein GpIb-alpha both of which have an intron in the 5-prime-noncoding region, a long second exon containing the complete coding region, and TATA-less promoters.Specifications
CD162 | |
Monoclonal | |
APC | |
SELPLG | |
CD162, CLA, PSGL-1, PSGL1, P-selectin glycoprotein ligand 1, cutaneous lymphocyte-associated associated antigen, SELPG, P-Selectin Glycoprotein Ligand 1 | |
Mouse | |
Purified | |
RUO | |
6404 | |
4° C | |
Liquid |
Flow Cytometry | |
TC2 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q14242 | |
SELPLG | |
Human CD162 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |