missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD162 (PSGL-1) Mouse anti-Human, PerCP-eFluor™ 710, Clone: FLEG, eBioscience™
Mouse Monoclonal Antibody
1052.00 SEK - 2760.00 SEK
Specifications
Antigen | CD162 (PSGL-1) |
---|---|
Clone | FLEG |
Target Species | Human |
Conjugate | PerCP-eFluor™ 710 |
Isotype | IgG2a, kappa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15569196
|
Affymetrix eBioscience
46-1629-41 |
25 Tests |
1052.00 SEK
25 tests |
25-02-2021 |
|||||
15579196
|
Affymetrix eBioscience
46-1629-42 |
100 Tests |
2760.00 SEK
100 tests |
25-02-2021 |
|||||
Description
Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor™ 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. SELPLG is the high affinity counter-receptor for P-selectin on myeloid cells and stimulated T lymphocytes. As such, it plays a critical role in the tethering of these cells to activated platelets or endothelia expressing P-selectin. The organization of the SELPG gene closely resembles that of CD43 and the human platelet glycoprotein GpIb-alpha both of which have an intron in the 5-prime-noncoding region, a long second exon containing the complete coding region, and TATA-less promoters.Specifications
CD162 (PSGL-1) | |
Human | |
IgG2a, kappa | |
Monoclonal | |
Liquid | |
Antibody | |
4° C, store in dark, DO NOT FREEZE! |
FLEG | |
PerCP-eFluor™ 710 | |
Flow Cytometry | |
Primary | |
Monoclonal | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
RUO |
For Research Use Only.