Learn More
CD162 (PSGL-1) Monoclonal Antibody (4RA10), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1942.00 SEK - 4590.00 SEK
Specifications
Antigen | CD162 (PSGL-1) |
---|---|
Clone | 4RA10 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16616404
|
Affymetrix eBioscience
53-1621-80 |
25 μg |
1942.00 SEK
Pack of 2 |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16626404
|
Affymetrix eBioscience
53-1621-82 |
100 μg |
4590.00 SEK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This 4RA10 monoclonal antibody reacts with mouse CD162, which is also known as P-selectin glycoprotein ligand-1 (PSGL-1). This clone has been successfully used in flow cytometry and has been reported to block binding of CD162 to CD62P and CD62L. Applications Reported: This 4RA10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4RA10 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
CD162 (PSGL-1) Monoclonal antibody specifically detects CD162 (PSGL-1) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD162 (PSGL-1) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
SELPLG | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
4RA10 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
Q62170 | |
20345 | |
Mouse PSGL-1 human IgG1 fusion protein | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
SELPLG |