Learn More
CD166 (ALCAM) Monoclonal Antibody (3A6), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
3395.00 SEK
Specifications
Antigen | CD166 (ALCAM) |
---|---|
Clone | 3A6 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15519206
|
Affymetrix eBioscience
46-1668-42 |
100 Tests |
3395.00 SEK
100 tests |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This 3A6 monoclonal antibody reacts with human CD166, which is also known as activated leukocyte cell adhesion molecule (ALCAM). CD166 is an immunoglobulin superfamily cell adhesion molecule that is broadly expressed on developing and adult hematopoietic (e.g., activated T and B cells, monocytes, and dendritic cells) and non-hematopoietic (e.g., neural, endothelial, and epithelial) tissues. CD166 has also been detected in melanoma, colorectal carcinoma, as well as cancers of the breast, prostate, and bladder. CD166 mediates cell-cell adhesion by forming low-affinity homophilic interactions (ALCAM-ALCAM) or high-affinity heterophilic interactions with its ligand CD6 (ALCAM-CD6). CD166 plays a role in development, inflammation, and neutrophil migration. Moreover, its expression can be used as a prognostic marker for several cancers and has been reported to be a cancer stem cell marker. The 3A6 antibody has been reported to induce the internalization of CD166 and its recycling back to the cell surface. Applications Reported: This 3A6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3A6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood lymphocytes. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
CD166 (ALCAM) Monoclonal antibody specifically detects CD166 (ALCAM) in Human samples. It is validated for Flow Cytometry.Specifications
CD166 (ALCAM) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q13740 | |
ALCAM | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ALCAM |
3A6 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
214 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.