missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD180 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30355.100uL
Additional Details : Weight : 0.02000kg
Description
Rabbit polyclonal antibody raised against partial recombinant human CD180.
- High specificity
- High reactivity
Specifications
CD180 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant human CD180. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
LY64/Ly78/MGC126233/MGC126234/RP105 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
4064 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
Q99467 | |
CD180 | |
Recombinant protein corresponding to human CD180. | |
100 μL | |
Primary | |
Human | |
Liquid |
For Research Use Only (RUO)