missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD184 (CXCR4) Monoclonal Antibody (2B11), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1716.00 SEK
Specifications
Antigen | CD184 (CXCR4) |
---|---|
Clone | 2B11 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16606334
|
Affymetrix eBioscience
25-9991-80 |
25 μg |
1716.00 SEK
Pack of 2 |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD184 (CXCR4) Monoclonal antibody specifically detects CD184 (CXCR4) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD184 (CXCR4) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
b2b220Clo; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD antigen CD184; CD184; CD184 antigen; chemokine (C-X-C motif) receptor 4; chemokine (C-X-C) receptor 4; chemokine receptor; chemokine receptor (LCR1); chemokine receptor 4; chemokine receptor CXCR4; Cmkar4; CXC; CXC chemokine receptor; C-X-C chemokine receptor type 4; CXC motif chemokine; C-X-C motif chemokine receptor 4; CXCR4; CXC-R4; CXCR-4; D2S201E; FB22; Fusin; HM89; HSY3RR; HUMSTR; LAP3; LAP-3; LCR1; LESTR; Leukocyte-derived seven transmembrane domain receptor; leukocyte-expressed seven-transmembrane-domain; lipopolysaccharide-associated protein 3; LPS-associated protein 3; neuropeptide Y receptor Y3; neuropeptide Y3 receptor; NPY3R; NPYR; NPYRL; NPYY3R; PB-CKR; PBSF/SDF-1; pre-B-cell-derived chemokine receptor; SDF-1 receptor; Sdf1r; seven transmembrane helix receptor; seven-transmembrane-segment receptor, spleen; stromal cell-derived factor 1 receptor; WHIM; WHIMS | |
Cxcr4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cxcr4 |
2B11 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
P70658 | |
12767 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |