missing translation for 'onlineSavingsMsg'
Learn More

CD185 (CXCR5) Mouse anti-Human, Non-human primate, Rhesus Monkey, APC, Clone: MU5UBEE, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9185-42

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568236

  • 3110.00 SEK / 100 tests


In stock, ready to ship: 5
Add to basket

Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. CXCR5 (C-X-Chemokine Receptor Type 5, CD185) also known as Burkitt lymphoma receptor 1 (BLR1) belongs to the CXC chemokine receptor family. CXCR5 is a G protein-coupled seven transmembrane receptor for chemokine BLC. CXCR5 gene is specifically expressed in Burkitt′s lymphoma and lymphatic tissues, such as follicles in lymph nodes, as well as in spleen. Studies of CXCR5 suggest that it plays an essential role in B cell migration. CXCR5 expression has been reported in human blood and salivary gland and in animal lymphatic organs, blood, bone marrow, brain, liver, lymph node, skin, spleen, stomach, and tonsil. ESTs (expressed sequence tags) have been isolated from human brain, germ cell, lung, lymph, and pancreas. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Research studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This "turn-off" activity in GPCRs have tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which are used in mental health and allergy applications, respectively.
Specifications

Specifications

CD185 (CXCR5)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P32302
CXC Chemokine Receptor 5, Fusin, LESTR, HUMSTR
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Non-human Primate, Rhesus Monkey
Flow Cytometry
MU5UBEE
APC
CXCR5
Liquid
CXCR5
IgG2b, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
643, 701792
Videos
SDS
Documents

For Research Use Only.