Learn More
CD19 Monoclonal Antibody (SJ25-C1), PE-Alexa Fluor™ 610, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1922
Additional Details : Weight : 0.50000kg
Description
The R-phycoerythrin (PE)-Cy7® tandem conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a 488 nm laser without occupying traditional channels associated with FITC and PE.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Monoclonal | |
0.3mg/mL | |
Assay-dependent | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Mouse, Mouse | |
purified | |
ASR | |
Primary | |
1 Each | |
Human | |
Liquid |
Flow Cytometry | |
SJ25-C1 | |
PE-Alexa Fluor 610 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P15391 | |
Cd19 | |
Nalm 1 Human cell line | |
500 μL | |
Kappa | |
930 | |
0.1% Sodium azide | |
4° C, store in dark | |
IgG1 κ |
Analyte Specific Reagent. Analytical and performance characteristics are not established.