missing translation for 'onlineSavingsMsg'
Learn More

CD196 (CCR6) Mouse anti-Human, APC, Clone: R6H1, eBioscience™

Mouse Monoclonal Antibody

1194.00 SEK - 2660.00 SEK

Specifications

Antigen CD196 (CCR6)
Clone R6H1
Target Species Human
Conjugate APC
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15568046
View Documents
Affymetrix eBioscience
17-1969-41
25 Tests
1194.00 SEK
25 tests
View alternative products
Save Up to 
»
Estimated Shipment
25-02-2021
Add to basket
 
15578046
View Documents
Affymetrix eBioscience
17-1969-42
100 Tests
2660.00 SEK
100 tests
View alternative products
Save Up to 
»
Estimated Shipment
25-02-2021
Add to basket
 
Description

Description

Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors. TRAIL/Apo2L is a new member of the TNF family and induces apoptosis of a variety of tumor cell lines. DR4 and DR5 are the recently identified functional receptors for TRAIL, and DcR1/TRID is a decoy receptor. Another member of the TRAIL receptor family was more recently identified and designated DcR2, TRAIL-R4, or TRUNDD. DcR2 has an extracellular TRAIL-binding domain but lacks intracellular death domain and does not induce apoptosis. Like DR4 and DR5, DcR2 transcript is widely expressed in normal human tissues. Overexpression of DcR2 attenuated TRAIL-induced apoptosis.
Specifications

Specifications

CD196 (CCR6)
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
R6H1
APC
Flow Cytometry
Primary
Monoclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.