missing translation for 'onlineSavingsMsg'
Learn More

CD1b Mouse anti-Human, APC, Clone: eBioSN13 (SN13 K5-1B8), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0018-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15567846

  • 1112.00 SEK / 25 tests


Estimated Shipment
22-02-2021
Add to basket

Description

Description

This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.
Specifications

Specifications

CD1b
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P29016
RP11-101J8.4, CD1, CD1A, MGC125990, MGC125991, R1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioSN13 (SN13 K5-1B8)
APC
CD1B
Liquid
CD1B
IgG1
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
910
Videos
SDS
Documents

For Research Use Only.