missing translation for 'onlineSavingsMsg'
Learn More

CD1b Mouse anti-Human, APC, Clone: eBioSN13 (SN13 K5-1B8), eBioscience™

Mouse Monoclonal Antibody

1112.00 SEK - 2490.00 SEK

Specifications

Antigen CD1b
Clone eBioSN13 (SN13 K5-1B8)
Target Species Human
Conjugate APC
Isotype IgG1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15577846
View Documents
Affymetrix eBioscience
17-0018-42
100 Tests
2490.00 SEK
100 tests
View alternative products
Save Up to 
»
Estimated Shipment
29-01-2021
Add to basket
 
15567846
View Documents
Affymetrix eBioscience
17-0018-41
25 Tests
1112.00 SEK
25 tests
View alternative products
Save Up to 
»
Estimated Shipment
25-02-2021
Add to basket
 
Description

Description

This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.
Specifications

Specifications

CD1b
Human
IgG1
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
eBioSN13 (SN13 K5-1B8)
APC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.