Learn More
CD1d Monoclonal Antibody (51.1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1112.00 SEK - 3440.00 SEK
Specifications
Antigen | CD1d |
---|---|
Clone | 51.1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15529056
|
Affymetrix eBioscience
46-0016-42 |
100 Tests |
3440.00 SEK
100 tests |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15519056
|
Affymetrix eBioscience
46-0016-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody 51.1 reacts with human CD1d, a member of the CD1 family with similarity to the non-polymorphic MHC Class I-like molecules. CD1d is a highly conserved single transmembrane receptor of the Immunoglobulin Superfamily. CD1d can associate with beta-microglobulin another feature showing similarity to MHC class I molecules, but can also exist as a nonglycosylated protein not in association with beta microglobulin. This suggests different control mechanisms for presenting glycolipid containing molecules to CD1d reactive NKT cells. Expression of CD1d is found on B cells of the periphery, in resting monocytes and cortical thymocytes. On intestinal epithelial cells (IEC) expression is polarized. Expression can also be found at low levels intracellularly in hepatocytes. In HCV (hepatitis C virus) livers, CD1d is highly expressed compared to normal controls. The 51.1 monoclonal antibody has been shown to have functional activity; blocking the interaction of CD1d transfected cells with NKT cells. Applications Reported: This 51.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 51.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
CD1d Monoclonal antibody specifically detects CD1d in Human samples. It is validated for Flow Cytometry.Specifications
CD1d | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P15813 | |
CD1D | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD1D |
51.1 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
912 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.