missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17336-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
CD1d is a 49 kDa heavy chain associated with Beta 2 microglobulin on cortical thymocytes. Beta 2 microglobulin independent expression of CD1d has also been demonstrated on human intestinal epithelial cells. CD1d is the sole group 2 member of the CD1 family of major histocompatibility (MHC) like glycoproteins. The CD1d gene encodes a divergent member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by the CD1d gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail. Diseases associated with CD1D include Mycobacterium Malmoense and Autoimmune Disease.Specifications
CD1d | |
Polyclonal | |
Unconjugated | |
CD1D | |
CD1A, CD1D, CD1d molecule, R3, R3G1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12479, 912 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P11609, P15813 | |
CD1D, Cd1d1 | |
CD1d Fusion Protein Ag11371 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |