missing translation for 'onlineSavingsMsg'
Learn More

CD1d, PerCP-eFluor™ 710, clone: 1B1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0011-80

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15579046

  • 1198.00 SEK / 25µg


Estimated Shipment
14-01-2021
Add to basket

Description

Description

Description: The 1B1 monoclonal antibody reacts with mouse CD1d, a 48kDa glycoprotein with structural homology to MHC class I molecules. While similar to MHC Class I, CD1d associates with beta2m, functionally CD1d is similar to MHC Class II. 1B1 detects CD1d at varying levels on mouse leukocytes. 1B1 detects beta2m associated CD1d.Applications Reported: This 1B1 antibody has been reported for use in flow cytometric analysis.Applications Tested: %This 1B1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.%.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD1d
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P11609
AI747460, CD1.1, Cd1a, Cd1d, Ly-38, T-cell surface glycoprotein CD1d1, antigen-presenting glycoprotein CD1d1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
1B1
PerCP-eFluor 710
Cd1d1
Liquid
Cd1d1
IgG2b, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12479
Videos
SDS
Documents

For Research Use Only.