missing translation for 'onlineSavingsMsg'
Learn More

CD200 Mouse anti-Human, Clone: 5F3D6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66282-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16838015

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD200 is a type-1 membrane glycoprotein that contains two immunoglobulin domains, and belongs to the immunoglobulin superfamily. CD200 is a broadly distributed cell surface glycoprotein that interacts with a receptor on myeloid cells (CD200R) and is implicated in locally preventing macrophage activation. Distant, but recognizable, homologues of CD200 have been identified in many herpesviruses and poxviruses. Studies of the related genes in mouse and rat suggest that CD200 may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms of CD200 have been found. Diseases associated with CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy.
Specifications

Specifications

CD200
Monoclonal
0.63 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P41217
CD200
CD200 Fusion Protein Ag5045
20 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
5F3D6
Unconjugated
CD200
CD200, CD200 molecule, MOX1, MOX2, MRC, OX 2, OX 2 membrane glycoprotein
Mouse
Protein G
RUO
4345
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers