missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
CD200/OX2 Antibody (OX-2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65320UV
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CD200/OX2 Monoclonal specifically detects CD200/OX2 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifikationer
| CD200/OX2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD200 | |
| Rat thymocyte membrane glycoproteins | |
| 0.25 mL | |
| Immunology | |
| 4345 | |
| Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| OX-2 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65320 recognizes the rat CD200 cell surface antigen, also known as OX-2, a type I membrane glycoprotein, expressed on rat thymocytes, neurons, follicular dendritic cells, endothelium and at low levels on some B cells and smooth muscle cells. Studies have indicated that the CD200-CD200 ligand system is important in the control of macrophage and granulocyte activation and that this antibody blocks OX-2 mediated T cell costimulation. This product is routinely tested by flow cytometry on rat thymocytes. | |
| Store at 4C in the dark. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering