missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 / BL-CAM Mouse, Clone: BLCAM/1795, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 933-MSM5-P0
Additional Details : Weight : 0.01000kg
Specifications
CD22 / BL-CAM | |
Cell Surface | |
BLCAM/1795 | |
Unconjugated | |
NP_001172030 | |
Mouse | |
130-140kDa | |
Protein A/G Purified | |
RUO | |
933 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD22 | |
Recombinant human BL-CAM/CD22 protein (aa52-178) | |
Raji, Daudi, IM9, JY25 and human peripheral blood lymphocytes. Human Tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |