Learn More
CD22 Monoclonal Antibody (eBio4KB128 (4KB128)), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-0229-41
Additional Details : Weight : 0.01000kg
Description
The eBio4KB128 MAb recognizes human CD22 (Siglec-2), which is a member of the siglec subgroup of the Ig superfamily. CD22 is a type I transmembrane glycoprotein composed of two polypeptide chains, CD22alpha and CD22beta, of 130 and 140 kDa respectively, produced by alternative splicing of the CD22 gene. CD22 is expressed at high levels on mature B cells and B cell lymphomas. The extracellular portion of CD22 contains seven Ig-like domains, some of which are capable of binding ligands with sialic acid moieties expressed on epithelial, endothelial, B and T cells. The intracellular portion of CD22 contains 6 tyrosine residues contained within immunotyrosine-based inhibitory motifs (ITIM) and immunotyrosine-based activation-like motifs, which are phosphorylated upon B-cell receptor engagement, which enables CD22 to participate in the positive and negative regulation of B-cell receptor signaling.
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Flow CytometrySpecifications
CD22 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P20273 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
eBio4KB128 (4KB128) | |
PE-Cyanine7 | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; Cd22; CD22 antigen; CD22 molecule; FLJ22814; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Siglec2; SIGLEC-2; T-cell surface antigen Leu-14 | |
Affinity chromatography | |
RUO | |
933 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |