missing translation for 'onlineSavingsMsg'
Learn More

CD223 (LAG-3) Mouse anti-Human, APC-eFluor® 780, Clone: 3DS223H, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-2239-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559576

  • 1374.00 SEK / 25 tests


Estimated Shipment
12-01-2021
Add to basket

Description

Description

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).
Specifications

Specifications

CD223 (Lag-3)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P18627
CD223, LAG-3, Ly66
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
3DS223H
APC-eFluor™ 780
LAG3
Liquid
LAG3
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3902
Videos
SDS
Documents

For Research Use Only.