Learn More
CD223 (LAG-3) Monoclonal Antibody (eBioC9B7W (C9B7W)), APC-eFluor™ 780, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1316.00 SEK - 3140.00 SEK
Specifications
Antigen | CD223 (LAG-3) |
---|---|
Clone | eBioC9B7W (C9B7W) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16340894
|
Affymetrix eBioscience
47-2231-80 |
25 μg |
1316.00 SEK
25µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16350894
|
Affymetrix eBioscience
47-2231-82 |
100 μg |
3140.00 SEK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The eBioC9B7W monoclonal antibody recognizes mouse CD223 (LAG-3, LAG3) protein expressed by activated alpha/beta-TCR bearing T cells, a subset of gamma/delta-TCR bearing T cells and a subset of NK cells. CD223 is a 70 kDa type I transmembrane protein with a structure that is similar to CD4. However, a soluble form of human CD223 has been detected by ELISA in human serum, and data suggest that mouse CD223 is proteolytically cleaved in the D4 domain. This results in a 54 kDa fragment containing all the extracellular domains, and a 16 kDa fragment containing the intracellular and transmembrane domains. The 54 kDa can remain membrane-associated or be released as soluble CD223. CD223 binds to MHC class II with higher affinity than CD4, and it is thought that this interaction is involved in the negative regulation of T-cell activation and homeostatic proliferation. Furthermore, CD223 is expressed by CD4+CD25+ regulatory T cells, and it has been suggested that CD223 may be involved in their regulatory function. This eBioC9B7W (C9B7W) antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD223 (LAG-3) Monoclonal antibody specifically detects CD223 (LAG-3) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD223 (LAG-3) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide | |
Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 | |
LAG3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
LAG3 |
eBioC9B7W (C9B7W) | |
Flow Cytometry | |
APC-eFluor 780 | |
Rat | |
Mouse | |
Q61790 | |
16768 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.