Learn More
Invitrogen™ CD226/DNAM-1 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHCD226X5
Additional Details : Weight : 0.33820kg
Description
The Human CD226/DNAM-1 ELISA quantitates Hu CD226 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CD226. Principle of the method The Human CD226 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes. This antibody is reported to inhibit T- and NK cell mediated cytotoxicity against tumour cell targets and to block TNF alpha and IFN gamma secretion by alloantigen-specific T-cells.Specifications
Q15762 | |
5 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
10666 | |
<12% | |
Biotinylated Detection Antibody, Adhesive Plate Covers, SAV-HRP, Assay Diluent Concentrate, Chromogen, Pre-coated 96 well plates, Wash Buffer, Standard, Stop Solution | |
5 x 96 Tests | |
Extracellular Matrix & Cell Adhesion | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
6.14 ng/mL to 600 ng/mL | |
5 ng/mL to 600 ng/mL | |
Biotin | |
CD Protein | |
This ELISA pair antibody detects human DNAM1. Other species not determined. | |
Human | |
Colorimetric Microplate Reader | |
DNAM-1,DNAM1,PTA1,TLiSA1 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
DNAM-1, DNAM1, PTA1, TLiSA1 | |
1 hr. 20 min. |