Learn More
CD235a Monoclonal Antibody (CLB-ery-1 (AME-1)), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHGLA04
Additional Details : Weight : 0.50000kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients. This antibody recognizes a papain-sensitive epitope in the extracellular domain of the sialoglycoprotein known as glycophorin A (CD235a).
CD235a Monoclonal antibody specifically detects CD235a in Human samples. It is validated for Flow Cytometry.Specifications
CD235a, Glycophorin A (CD235a) | |
Monoclonal | |
PE | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Mouse | |
purified | |
ASR | |
2993 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
CLB-ery-1 (AME-1) | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
P02724 | |
GYPA | |
Human Glycophorin A (CD235a) | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |