missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), APC-eFluor™ 780, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

1816.00 SEK - 4315.00 SEK

Specifikationer

Antigen CD24
Klona eBioSN3 (SN3 A5-2H10)
Koncentration 5 μL/Test
Användningsområden Flow Cytometry
Klassificering Monoclonal
Se fler specifikationer

Produkter 2
Produktkod Brand Kvantitet Pris Kvantitet och tillgänglighet  
Produktkod Brand Kvantitet Pris Kvantitet och tillgänglighet  
15589526
Visa dokument
Invitrogen™
47-0247-41
25 Tests
1816.00 SEK
25 tester
Se alternativa produkter
Spara upp till 
»
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
15599526
Visa dokument
Invitrogen™
47-0247-42
100 Tests
4315.00 SEK
100 tester
Se alternativa produkter
Spara upp till 
»
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning

Beskrivning

Description: The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50 kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24. Applications Reported: This eBioSN3 (SN3 A5-2H10) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioSN3 (SN3 A5-2H10) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifikationer

Specifikationer

CD24
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen
Cd24
Primary
4°C, store in dark, DO NOT FREEZE!
Cd24
eBioSN3 (SN3 A5-2H10)
Flow Cytometry
APC-eFluor 780
Mouse
Human
P25063
100133941
IgG1 κ
Affinity chromatography
Antibody
Filmer
SDS
Dokument

Dokument

Produktspecifikationer

For Research Use Only.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.