missing translation for 'onlineSavingsMsg'
Learn More

CD24 Mouse anti-Human, PerCP-eFluor™ 710, Clone: eBioSN3 (SN3 A5-2H10), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0247-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15579096

  • 1210.00 SEK / 25 tests


Estimated Shipment
11-01-2021
Add to basket

Description

Description

00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications

Specifications

CD24
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P25063
CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24A; FLJ22950; FLJ43543; MGC75043
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioSN3 (SN3 A5-2H10)
PerCP-eFluor™ 710
CD24
Liquid
CD24
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
100133941
Videos
SDS
Documents

For Research Use Only.