Learn More
CD25 Monoclonal Antibody (BC96), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1658.00 SEK - 3565.00 SEK
Specifications
Antigen | CD25 |
---|---|
Clone | BC96 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15855723
|
Affymetrix eBioscience
67-0259-42 |
100 Tests |
3565.00 SEK
Pack of 100 |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15845723
|
Affymetrix eBioscience
67-0259-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The LG.7F9 monoclonal antibody reacts with mouse CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by virtually all mature T cells and by a subpopulation of B cells, mainly memory B cells. In mouse, CD27 has been found on nearly all thymocytes excluding a population of CD46-CD8- precursors. CD27 binds to CD70 and, through this interaction, plays an important role in T cell-B cell interaction. It has been reported that triggering CD27 plays an important role in the maturation of CD4+ and CD8+ effector cells. LG.7F9 cross-reacts with human and rat CD27. Applications Reported: This LG.7F9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This LG.7F9 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.
CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for Flow Cytometry.Specifications
CD25 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P01589 | |
IL2RA | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL2RA |
BC96 | |
Flow Cytometry | |
Super Bright 702 | |
Mouse | |
Human | |
3559 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |