missing translation for 'onlineSavingsMsg'
Läs mer

CD27 Monoclonal Antibody (LG.7F9), Functional Grade, eBioscience™, Invitrogen™

Produktkod. 17180962
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17180962 25 mg
17190962 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17180962 Leverantör Affymetrix eBioscience Leverantörsnummer 160271025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Armenian Hamster Monoclonal Antibody

The LG.7F9 MAb reacts with mouse CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by virtually all mature T cells and by a subpopulation of B cells, mainly memory B cells. In mouse, CD27 has been found on nearly all thymocytes excluding a population of CD46-CD8- precursors. CD27 binds to CD70 and, through this interaction, plays an important role in T cell-B cell interaction. It has been reported that triggering CD27 plays an important role in the maturation of CD4+ and CD8+ effector cells. LG.7F9 cross-reacts with human and rat CD27. Applications Reported: The LG.7F9 antibody has been reported for use in flow cytometric analysis. LG.7F9 has also been reported in blocking of ligand binding. Applications Tested: The LG.7F9 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

CD27 Monoclonal antibody specifically detects CD27 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen CD27
Användningsområden Flow Cytometry, Functional Assay, Neutralization
Klassificering Monoclonal
Klona LG.7F9
Koncentration 1 mg/mL
Konjugera Functional Grade
Formulering PBS with no preservative; pH 7.2
Gen CD27
Genaccessionsnr. P26842, P41272
Gene Alias CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; Tnfrsf7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7
Värd art Armenian Hamster
Reningsmetod Affinity chromatography
Kvantitet 25 mg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 21940, 500318, 939
Målarter Human, Mouse, Rat
Innehåll och lagring 4° C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.