Learn More
CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1534.00 SEK - 3715.00 SEK
Specifications
Antigen | CD274 (PD-L1, B7-H1) |
---|---|
Clone | MIH5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15549326
|
Affymetrix eBioscience
46-5982-80 |
25 μg |
1534.00 SEK
25µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15559326
|
Affymetrix eBioscience
46-5982-82 |
100 μg |
3715.00 SEK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody. Applications Reported: This MIH5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MIH5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD274 (PD-L1, B7-H1) Monoclonal antibody specifically detects CD274 (PD-L1, B7-H1) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD274 (PD-L1, B7-H1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
Cd274 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd274 |
MIH5 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Mouse | |
Q9EP73 | |
60533 | |
IgG2a λ | |
Affinity chromatography | |
Antibody |
For Research Use Only.