missing translation for 'onlineSavingsMsg'
Läs mer

CD274, NA/LE Mouse anti-Human, Unlabeled, Clone: MIH1, BD

Produktkod. 15874629
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
15874629 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15874629 Leverantör BD Biosciences Leverantörsnummer 565959

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

The MIH1 monoclonal antibody specifically binds to CD274, which is also known as, B7 homolog 1 (B7-H1), Programmed cell death 1 ligand 1 (PDCD1 ligand, PDCD1L1, PDCD1LG1), or Programmed death ligand 1 (PD-L1, PDL1). CD274 and PD-L2 (CD273) are type I transmembrane glycoproteins that belong to the B7 family and serve as ligands for CD279 (Program Death 1/PD-1). CD274 is expressed on antigen-presenting cells including activated monocytes/macrophages and dendritic cells, as well as, activated T cells, and keratinocytes. CD274 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, and on most carcinomas. CD274 plays an important role in regulating T cell responses. The MIH1 antibody blocks CD279 binding to CD274 and can enhance the proliferation and cytokine production of activated T cells.

NA/LE
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen CD274, NA/LE
Klassificering Monoclonal
Klona MIH1
Koncentration 1.0mg/mL
Konjugera Unconjugated
Formulering No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Gene Alias B7-H1, B7-H, PD-L1, PDL1, PDCD1 ligand 1, PDCD1L1, PDCD1LG1
Värd art Mouse
Immunogen Human CD274 Transfected Cell Line
Reningsmetod Affinity Purified
Kvantitet 500 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Målarter Human
Innehåll och lagring Store undiluted at 4°C.
Isotyp IgG1 κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.