missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD278 (ICOS) Monoclonal Antibody (ISA-3), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1258.00 SEK - 3030.00 SEK
Specifications
Antigen | CD278 (ICOS) |
---|---|
Clone | ISA-3 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16626454
|
Affymetrix eBioscience
56-9948-42 |
100 Tests |
3030.00 SEK
100 tests |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16616454
|
Affymetrix eBioscience
56-9948-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CD278 (ICOS) Monoclonal antibody specifically detects CD278 (ICOS) in Human samples. It is validated for Flow Cytometry.Specifications
CD278 (ICOS) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
29851 | |
IgG1 κ | |
Affinity chromatography | |
ICOS |
ISA-3 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
Human | |
Q9Y6W8 | |
ICOS | |
Primary | |
Antibody |