Läs mer
Invitrogen™ CD281 (TLR1) Monoclonal Antibody (eBioTR23 (TR23)), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
16850.00 SEK - 38090.00 SEK
Specifikationer
| Antigen | CD281 (TLR1) |
|---|---|
| Klona | eBioTR23 (TR23) |
| Koncentration | 0.5 mg/mL |
| Användningsområden | Flow Cytometry, Immunoprecipitation |
| Klassificering | Monoclonal |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
17180692
|
Invitrogen™
14-9011-37 |
2 mg |
16850.00 SEK
2mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
17190692
|
Affymetrix eBioscience
14-9011-95 |
10 mg | |||||||
Beskrivning
Description: The eBioTR23 monoclonal antibody reacts with mouse Toll-like receptor 1 (TLR1, CD281). TLR1 is an approximately 98 kDa protein belonging to the Toll-like receptor family, and is closely-related to mouse TLR6 in both structure and function. Through the formation of a complex with TLR2, TLR1 is able to bind bacterially-derived triacyl lipopeptides, as well as soluble factors released from Neisseria meningitidis. TLR1 is expressed by macrophages and macrophages in TLR1-deficient mice display impaired production of inflammatory cytokines upon stimulation with triacyl lipopeptides and lipoproteins from mycobacteria. Applications Reported: This eBioTR23 (TR23) antibody has been reported for use in flow cytometric analysis, and immunoprecipitation. Applications Tested: This eBioTR23 (TR23) antibody has been tested by flow cytometric analysis of mouse TLR1-transfected Baf/3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
TLR1 protein is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. The Toll-like receptor (TLR) family in mammals are comprised of a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and an IL-1 receptor motif in the cytoplasmic domain. The TLRs could constitute an important and unrecognized component of innate immunity in humans. In mice, TLR1 interacts with TLR2 and co-expression of TLR1 and TLR2 enhance the NF-kappaB activation in response to a synthetic lipopeptide. They recognize the lipid configuration of the native mycobacterial lipoprotein as well as several tri-acylated lipopeptides. Different length transcripts presumably resulting from use of alternative polyadenylation site, and/or from alternative splicing, have been noted for the TLR1 gene.Specifikationer
| CD281 (TLR1) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 21897 | |
| Primary | |
| 4°C | |
| TLR1 |
| eBioTR23 (TR23) | |
| Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9EPQ1 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.