Learn More
CD29 (Integrin beta 1) Monoclonal Antibody (TS2/16), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1484.00 SEK - 3440.00 SEK
Specifications
Antigen | CD29 (Integrin beta 1) |
---|---|
Clone | TS2/16 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933724
|
Affymetrix eBioscience
67-0299-42 |
100 Tests |
3440.00 SEK
Pack of 100 |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923724
|
Affymetrix eBioscience
67-0299-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This monoclonal antibody 9A11-CD33 reacts with mouse CD33, also known as Siglec3 and gp67. CD33 belongs to the sialic acid-binding immunoglobulin lectins (Siglec) family that binds to sialic acid-containing glycans on cell surface glycoproteins and glycolipids on mammalian cells. Mouse CD33 binds to short O-linked glycans of certain mucins and sialyl-Tn epitopes. Mouse CD33 is expressed by mature granulocytes, immature myeloid cells, and microglia. Unlike human CD33, mouse CD33 lacks a cytoplasmic ITIM domain and exists as two splice variants that have different cytoplasmic tails. In microglia, CD33 inhibits the phagocytosis of toxic amyloid beta. Applications Reported: This 9A11-CD33 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9A11-CD33 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter.
CD29 (Integrin beta 1) Monoclonal antibody specifically detects CD29 (Integrin beta 1) in Human samples. It is validated for Flow Cytometry.Specifications
CD29 (Integrin beta 1) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P05556 | |
ITGB1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ITGB1 |
TS2/16 | |
Flow Cytometry | |
Super Bright 702 | |
Mouse | |
Human | |
3688 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |