missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd2ap Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24122-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.Specifications
Cd2ap | |
Polyclonal | |
Unconjugated | |
CD2AP | |
AL024079, C78928, CD2 associated protein, Cd2ap, METS 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23607 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y5K6 | |
CD2AP | |
Cd2ap Fusion Protein Ag21170 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |