missing translation for 'onlineSavingsMsg'
Learn More

CD3 delta Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16669-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826374

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The encoded membrane protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in this gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms have been found for this gene. Other variants may also exist, but the full-length natures of their transcripts has yet to be defined.
Specifications

Specifications

CD3 delta
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04234, P04235
CD3 delta, CD3D, CD3-delta, T cell receptor T3 delta chain, T3D
Rabbit
IgG
150 μL
-20°C
Primary
12500, 915
Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
CD3D
Liquid
CD3D
CD3 delta Fusion Protein Ag10148
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers