Learn More
IBA Lifesciences CD3 Isolation Kit for FABian™
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit, Human
10 Kits, Human
Unit Size:
1 set
10 sets
Description
- The kit is used for the selection of CD3+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
- It includes CD3 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
- Each kit is sufficient for one FABian™ run to isolate cells from 9mL whole blood.
Specifications
Specifications
Product Type | Non-magnetic Cell Isolation Kit |
Content And Storage | 2°C to 8°C |
Donor Source | Human Whole Blood |
For Use With (Application) | Isolation of label-free cells |
Sufficient For | 9 mL Whole Blood |
Quantity | 10 Kits, Human |
Product Content Correction
Product Title
IBA Lifesciences CD3 Isolation Kit for FABian™ >
Spot an opportunity for improvement?Share a Content Correction